Skip to Content
Merck
All Photos(2)

Key Documents

MAB10422

Sigma-Aldrich

Anti-Plexin-A3 Antibody, clone 18A9.2

clone 18A9.2, from mouse

Synonym(s):

plexin A3, PLEXIN-A3, plexin-A3, Sex chromosome X transmembrane protein of HGF receptor family 3, Semaphorin receptor SEX, plexin-4, Plexin-4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€379.00

€379.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€379.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€379.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

18A9.2, monoclonal

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PLXNA3(55558)

General description

Plexin-A3 is a cell surface receptor for semaphorin that controls signaling for neuronal axon regulation. Plexin-A3 is thought to play a role in Sema3F and Sema3A signaling, has been described as essential for dorsal turning of the corticospinal tract axons and may be an important contributor to development of hippocampal axonal projections. Plexin-A3 has been found expressed in newly-differentiated sympathetic neurons, but not their neural crest precursors, and together with Plexin-A4, functions cooperatively to regulate the migration of sympathetic neurons and then differentially to guide the sympathetic axons.

Immunogen

GST-tagged recombinant protein corresponding to human Plexin-A3.

Application

Anti-Plexin-A3 Antibody, clone 18A9.2 is a Mouse Monoclonal Antibody for detection of Plexin-A3 also known as PLEXIN-A3, Semaphorin receptor SEX or Plexin-4 & has been validated in WB & IHC.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
Western Blot Analysis: 1 µg/mL from a previous lot detected Plexin-A3 on 10 µg of HT-29 and NIH/3T3 cell lysates.

Immunohistochemistry Analysis: 1:400 dilution from a previous lot detected Plexin-A3 in colorectal carcinoma tissue.

Quality

Evaluated by Western Blot in PC-12 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Plexin-A3 on 10 µg of PC-12 cell lysate.

Target description

~220 kDa observed. The calculated molecular weight of this protein is 208 kDa; however, a higher band may be observed due to glycosolation.

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
PC-12 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service