IEVH07804
Immobilon®-E PVDF Membrane
4 sheets, 7 cm x 8.4 cm, The first PVDF transfer Membrane for western blotting that wets out in transfer buffer or water. For chemiluminescence or chromogenic detection.
About This Item
Recommended Products
material
PVDF membrane
plain filter
white filter
Quality Level
feature
easily wetted with water
hydrophobic
manufacturer/tradename
Immobilon®
technique(s)
western blot: suitable
filter L × W
7 cm × 8.4 cm
pore size
0.45 μm pore size
compatibility
for use with Amido black
for use with Colloidal gold
for use with Coomassie brilliant blue
for use with India ink
for use with Ponceau-S red
for use with Sypro<TMSYMBOL></TMSYMBOL> ruby
for use with Transillumination
detection method
chemiluminescent
colorimetric
radioactive
shipped in
ambient
General description
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service