Skip to Content
Merck
All Photos(1)

Documents

GSWP29325

Millipore

MF-Millipore Membrane

pore size 0.22 μm, filter diam. 293 mm, hydrophilic

Synonym(s):

MF-Millipore Membrane, mixed cellulose esters, Hydrophilic, 0.22 μm, 293 mm, white, plain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602

material

mixed cellulose esters (MCE) membrane
plain filter
white filter

Quality Level

feature

hydrophilic

manufacturer/tradename

MF-Millipore

parameter

18 mL/min-cm2 water flow rate
2 L/min-cm2 air flow rate
75 °C max. temp.

filter diam.

293 mm

thickness

180 μm

gravimetric extractables

2%

refractive index

n/D 1.51

matrix

MF-Millipore

pore size

0.22 μm pore size
75 % porosity

bubble point

≥3.52 bar, air with water at 23 °C

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service