GF026
Granulocyte-Macrophage Colony-Stimulating Factor Protein, Recombinant mouse
The Granulocyte Macrophage Colony Stimulating Factor protein (GM-CSF protein) is a potent species specific stimulator of bone marrow cells.
Synonym(s):
GM-CSF
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
eCl@ss:
32160405
Recommended Products
biological source
mouse
Assay
>98% (SDS-PAGE and HPLC)
specific activity
≥5 x 106 U/mg
manufacturer/tradename
Chemicon®
technique(s)
cell culture | mammalian: suitable
input
sample type hematopoietic stem cell(s)
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Gene Information
mouse ... IL3(16187)
General description
Granulocyte Macrophage Colony Stimulating Factor (GM-CSF) is a potent species specific stimulator of bone marrow cells. GM-CSF stimulated precursor cells of granulocytes, macrophages, and eosinophils. Murine GM-CSF is a 14.0 kDa protein containing 124 amino acid residues.
Application
For most in vitro applications, murine GM-CSF exerts its biological activity in the concentration range of 0.05 to 0.5 ng/mL. Responding cells are (partial list): Certain BM precursor cells
Physical form
Sterile filtered solution lyophilized from 10mM HoAc.
Storage and Stability
The lyophilized murine GM-CSF, though stable at room temperature, is best stored desiccated below 0°C. Reconstitute in water to a concentration of 100 ng/uL. This solution can be diluted into other buffered solutions. Reconstituted murine GM-CSF should be stored in working aliquots at -20°C.
Analysis Note
Specific Activity: Chemicon′s mouse GM-CSF is fully biologically active when compared to standards.The ED50 as determined by the dose-dependent stimulation of the proliferation of mouse FDC-P1 cells in ≤ 0.2 ng/mL
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jingyue Yan et al.
Advanced materials (Deerfield Beach, Fla.), 34(47), e2207486-e2207486 (2022-09-20)
Toll-like receptors (TLRs) and CD40-related signaling pathways represent critical bridges between innate and adaptive immune responses. Here, an immunotherapy regimen that enables co-stimulation of TLR7/8- and CD40-mediated pathways is developed. TLR7/8 agonist resiquimod (R848) derived amino lipids, RAL1 and RAL2, are
Annadurai Thangaraj et al.
Redox biology, 40, 101843-101843 (2021-01-02)
The advent of combined antiretroviral treatment (cART) as a treatment for HIV-1 infection has not only resulted in a dramatic decrease in the peripheral viral load but has also led to increased life expectancy of the infected individuals. Paradoxically, increased
Annadurai Thangaraj et al.
Autophagy, 16(2), 289-312 (2019-04-17)
Although cocaine exposure has been shown to potentiate neuroinflammation by upregulating glial activation in the brain, the role of mitophagy in this process remains an enigma. In the present study, we sought to examine the role of impaired mitophagy in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service