Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB191F

Sigma-Aldrich

Milli-Mark® Anti-CD27-FITC Antibody, clone M-T271

clone M-T271, Milli-Mark®, from mouse

Synonym(s):

CD27 antigen, CD27L receptor, T-cell activation antigen CD27, T14, Tumor necrosis factor receptor superfamily member 7

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
€370.00

€370.00


Estimated to ship onMay 16, 2025


Request a Bulk Order

Select a Size

Change View
100 TESTS
€370.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

€370.00


Estimated to ship onMay 16, 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

M-T271, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD27(939)

General description

CD27, a member of the nerve growth factor receptor (NGFR) superfamily, is a type I transmembrane protein consisting of a disulphide-linked 120 kDa homodimer (Bigler, 1988; Sugita, 1992; Morimoto, 1995). CD27 is expressed on mature thymocytes and on the majority of human peripheral blood T cells, on both CD4+ and CD8+ subsets. CD27 is also expressed on activated B cells and a fraction of resting natural killer (NK) cells (Maurer, 1990; Sugita, 1992). Among CD4+ T cells, CD27 is preferentially expressed on unprimed CD4+CD45RA+CD45R0- T cells, while primed CD4+CD45RA-CD45R0+ T cells express low levels of CD27 (Sugita, 1992). During activation, the expression of CD27 is increased on B cells and unprimed T cells, while primed T cells have a relatively low capacity for upregulation of CD27 (Morimoto, 1995; Hintzen, 1994). The activation is also accompanied by the appearance of a 32 kDa soluble form of CD27 (sCD27) found in supernatants of activated lymphocytes (Hintzen, 1994).
CD27 is believed to play a role in T cell activation, since the expression is upregulated during T cell activation (Bigler, 1988; Sugita, 1992; Morimoto, 1995; Maurer, 1990; Hintzen, 1994; Lens, 1995).

Specificity

Antibody recognizes Human CD27 antigen.

Immunogen

Leukemic peripheral blood T cells from a patient with chronic T cell leukemia (T-CLL)

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Milli-Mark Anti-CD27-FITC Antibody, clone M-T271 is validated for use in FC for the detection of CD27.

Quality

Evaluated by flow cytometry using human PBMCs

Target description

120 kDa (Bigler, 1988; Sugita, 1992; Morimoto, 1995)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintained refrigerated at 2-8°C from date of receipt. Protected from light in undiluted aliquots for 6 months from date of receipt.

Analysis Note

Control
PBMCs

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service