Skip to Content
Merck
All Photos(1)

Key Documents

ABT336

Sigma-Aldrich

Anti-BCAR3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Breast cancer anti-estrogen resistance 3, NSP2, SH2D3B, Novel SH2-containing protein 2, SH2 domain-containing protein 3B, BCAR

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€326.00

€326.00


Estimated to ship onMay 07, 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€326.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€326.00


Estimated to ship onMay 07, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

human (immunogen homology)

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BCAR3(8412)

General description

Breast cancer anti-estrogen resistance protein 3 (BCAR3), also known as Novel SH2-containing protein 2 (NSP2) and SH2 domain-containing protein 3B (SH2D3B) is an adapter protein that regulates the proliferation in breast cancer cells. BCAR3 may also be regulated by cellular adhesion to extracellular matrix proteins. BCAR3 is found in several cancer cell lines; however, it is not found in nonmalignant breast tissue. When BCAR3 is overexpressed, it confers anti-estrogen resistance in breast cancer cell lines.

Immunogen

KLH-conjugated linear peptide corresponding to human BCAR3.

Application

Anti-BCAR3 Antibody is a highly specific rabbit polyclonal antibody, that targets Breast cancer Anti-estrogen resistance protein 3 & has been tested in western blotting.
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in mouse kidney tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected BCAR3 in 15 µg of mouse kidney tissue lysate.

Target description

~104 kDa observed

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse kidney tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service