Skip to Content
Merck
All Photos(3)

Documents

ABN1486

Sigma-Aldrich

Anti-PDE2A

from rabbit, purified by affinity chromatography

Synonym(s):

cGMP-dependent 3′, 5′-cyclic, phosphodiesterase, PDE2A, Cyclic GMP-stimulated phosphodiesterase, cGSPDE, CGS-PDE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PDE2A(5138)

General description

cGMP-dependent 3′,5′-cyclic phosphodiesterase (UniProt: O00408; also known as EC:3.1.4.17, PDE2A, Cyclic GMP-stimulated phosphodiesterase, CGS-PDE, cGSPDE) is encoded by the PDE2A gene ( Gene ID: 5138) in human. PDE2A is an evolutionarily conserved cyclic nucleotide phosphodiesterase with a dual-specificity for the second messengers cAMP and cGMP that are key regulators of many important physiological processes. PDE2A is expressed in both the periphery and in the central nervous system, however its expression is highest in the brain, where it is mainly localized in the cortex, hippocampus, and striatum. Three splice variants of PDE2A have been described, the PDE2A1, PDE2A2 and PDE2A3. PDE2A1 is cytosolic whereas -A2 and -A3 are membrane bound. PDE2A isoform 2 is specifically targeted to the mitochondrial matrix, where it forms a local signaling system with soluble adenylate cyclase regulating the respiratory chain. PDE2A is a dimeric enzyme that binds 2 divalent metal cations per subunit. Site 1 may preferentially bind zinc ions, while site 2 has a preference for magnesium and/or manganese ions. Each PDE2A monomer contains an N-terminal (200 residues) domain of unknown function, tandem GAF domains (GAF A and GAF B), and a C-terminal catalytic domain. GAF A functions as a dimerization locus. PDE2 A catalytic activity is allosterically stimulated by cGMP binding to its GAF B domain. Binding of cGMP induces conformational change in the protein structure leading to higher enzyme activity. Ref.: Acin-Perez, R., et al. (2011). J. Biol. Chem. 286 (35): 30423-30432; Martinez, SE., et al. (2002). Proc. Natl. Acad. Sci. USA 99(20); 13260-13265; Trabanco, A., et al. (2016). Expert Opin. Therap. Patents 26(8): 933-946).

Specificity

This polyclonal antibody specificaaly detects PDE2A in human.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the N-terminal region of human PDE2A.

Application

Anti-PDE2A, Cat. No. ABN1486, is a highly specific rabbit polyclonal antibody that targets cGMP-dependent 3′,5′-cyclic phosphodiesterase and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000 dilution of this antibody detected PDE2A in human cerebellum, human cerebral cortex, human skin, human lung, human kidney, and human heart tissues.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in human cerebral cortex tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected PDE2A in 10 µg of human cerebral cortex tissue lysate.

Target description

~106 kDa observed; 105.72 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service