Skip to Content
Merck
All Photos(2)

Key Documents

ABE344

Sigma-Aldrich

Anti-Acetyl MLL1 (KMT2A) (Lys 1130/1133)

from rabbit

Synonym(s):

Histone-lysine N-methyltransferase 2A, EC: 2.1.1.43, Lysine N-methyltransferase 2A, ALL-1, CXXC-type zinc finger protein 7, Myeloid/lymphoid or mixed-lineage leukemia, Myeloid/lymphoid or mixed-lineage leukemia protein 1, Trithorax-like protein, Zinc fin

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€130.00
100 μG
€341.00

€130.00


Estimated to ship onMay 09, 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€130.00
100 μG
€341.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€130.00


Estimated to ship onMay 09, 2025


Request a Bulk Order

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

dot blot: suitable
inhibition assay: suitable (peptide)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

acetylation (Lys1130/Lys1133)

Gene Information

human ... KMT2A(4297)

Related Categories

General description

Histone-lysine N-methyltransferase 2A (UniProt: Q03164; also known as EC: 2.1.1.43, Lysine N-methyltransferase 2A, ALL-1, CXXC-type zinc finger protein 7, Myeloid/lymphoid or mixed-lineage leukemia, Myeloid/lymphoid or mixed-lineage leukemia protein 1, MLL1, Trithorax-like protein, Zinc finger protein HRX) is encoded by the KMT2A (also known as ALL1, CXXC7, HRX, HTRX, MLL, MLL1, TRX1) gene (Gene ID: 4297) in human. It is expressed in heart, lung, and brain and in T- and B-lymphocytes and plays an essential role in early development and hematopoiesis. Proteolytic cleavage of MLL1 by Taspase 1 (TASP1) generates an N-terminal cleavage product of 320 kDa and a C-terminal product of 180 kDa. The catalytic subunit of the MLL1/MLL complex, a multiprotein complex that mediates both methylation of Lysine 4 of histone H3 (H3K4me) complex and acetylation of Lysine 16 of histone H4 (H4K16ac). In the MLL1/MLL complex, it specifically mediates H3K4me, a specific tag for epigenetic transcriptional activation. MLL1 displays weak methyltransferase activity by itself, and requires other component of the MLL1/MLL complex to obtain full methyltransferase activity. It does not exhibit any activity toward histone H3 phosphorylated on Threonine 3, and less activity toward H3 dimethylated on Arginine 8 or Lysine 9. However, it has higher activity toward H3 acetylated on Lysine 9. MLL1 is shown to be required for transcriptional activation of HOXA9 and it promotes PPP1R15A-induced apoptosis. MLL1 is reported to coordinate recruitment of CLOCK BMAL1 activator complexes to chromatin, an event associated with cyclic trimethylation of histone H3 Lysine 4 (H3K4) at circadian promoters. MLL1 is also reported to play a critical role in the control of circadian gene expression and is essential for the transcriptional activation mediated by the CLOCK-ARNTL/BMAL1 heterodimer. (Ref.: Aguilar-Arnal, L., et al. (2015). Nat. Struct. Mol. Biol. 22(4); 312-318).

Specificity

This rabbit polyclonal antibody specifically detects human Histone-lysine N-methyltransferase 2A acetylated on lysine 1130 and 1133.

Immunogen

Epitope: unknown
KLH-cpnugated linear peptide corresponding to 12 amino acids surrounding acetylated lysines 1130 and 1133 from the N-terminal half of human Histone-lysine N-methyltransferase 2A.

Application

Anti-Acetyl MLL1 (KMT2A) (Lys 1130/1133), Cat. No. ABE344, is a highly specific rabbit polyclonal antibody that targets Histone-lysine N-methyltransferase 2A (MLL1) acetylated on Lysine 1130/1133 and has been tested for use in Dot Blot, Peptide Inhibition Assay, and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A representative lot detected Acetyl MLL1 (KMT2A) (Lys 1130/1133) in Western Blotting applications (Aquilar-Arnal, L., et. al. (2015). Nat Struct Mol Biol. 22(4):312-8).

Peptide Inhibition Analysis: A representative lot was used with Acetylated peptide (Ag peptide) and Control peptide for peptide block analysis. (Courtesy of Dr. Lorena Aguilar-Arnal)

Quality

Evaluated by Dot Blot with Acetylated peptide (Ag peptide) and Control peptide.



Dot Blot Analysis: 1 µg/mL of this antibody detected Acetyl MLL1 (KMT2A) peptide acetylated on Lysine 1130/1133, but did not dectect the control peptide.


Target description

431.76 kDa calculated.

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl, with 0.05% Sodium Azide without glycerol.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service