Skip to Content
Merck
All Photos(1)

Key Documents

ABE1317

Sigma-Aldrich

Anti-phospho Cyclin-dependent kinase 7 (Thr170) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Cyclin-dependent kinase 7, 39 kDa protein kinase, p39 Mo15, CDK-activating kinase 1, Cell division protein kinase 7, Serine/threonine-protein kinase 1, TFIIH basal transcription factor complex kinase subunit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€381.00

€381.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€381.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€381.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

technique(s)

ChIP: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr170)

Gene Information

human ... CDK7(1022)

General description

Cyclin-dependent kinases (CDKs) are activated by the binding to a cyclin protein and they mediate the progression through the cell cycle. Each different complex controls a specific transition between 2 subsequent phases in the cell cycle. Cyclin-dependent kinase 7 (CDK7), also known as Cyclin-dependent kinase 7, or 39 kDa protein kinase (p39-Mo15), CDK-activating kinase 1, Cell division protein kinase 7, Serine/threonine-protein kinase 1, or TFIIH basal transcription factor complex kinase subunit, and encoded by the gene CDK7/CAK/CAK1/CDKN7/MO15/STK1, is a serine/threonine kinase involved in cell cycle control and in RNA polymerase II-mediated RNA transcription. CDK7 is the catalytic subunit of the CDK-activating kinase (CAK) complex. CAK activates the cyclin-associated kinases CDK1, CDK2, CDK4 and CDK6 by threonine phosphorylation, thus regulating cell cycle progression. CDK7 is inactivated by phosphorylation, thus Cyclin-dependent kinase 7 (Thr107) is an inactive CDK7 protein.

Immunogen

Epitope: Phosphorylated T107
Linear peptide corresponding to human phosphorylated phospho Cyclin-dependent kinase 7 (Thr170).

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-phospho Cyclin-dependent kinase 7 (Thr170) antibody is validated for use in western blotting & ChIP for the detection of phospho Cyclin-dependent kinase 7 (Thr170).
Western Blotting Analysis: A representative lot detected phospho Cyclin-dependent kinase 7 (Thr170) in Cdk7 and Csk1 kinase (Larochelle, S., et al. (2012). Nature Struc. Mole. Biol. 19(11):1108-1115).
Chromatin Immunoprecipitation Analysis: A representative lot immunoprecipitated phospho Cyclin-dependent kinase 7 (Thr170) in HCT116 cell lysate treated with doxorubicin (Larochelle, S., et al. (2012). Nature Struc. Mole. Biol. 19(11):1108-1115).

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected phospho Cyclin-dependent kinase 7 (Thr170) in 10 µg of A431 cell lysate.

Target description

~ 39 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service