Skip to Content
Merck
All Photos(1)

Key Documents

ABE12

Sigma-Aldrich

Anti-phospho CARM1 (Ser228) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Protein arginine N-methyltransferase 4, coactivator-associated arginine methyltransferase 1, coactivator-associated arginine methyltransferase-1

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
€344.00

€344.00


Estimated to ship onApril 23, 2025


Request a Bulk Order

Select a Size

Change View
200 μG
€344.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€344.00


Estimated to ship onApril 23, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, horse

species reactivity (predicted by homology)

canine (based on 100% sequence homology), equine (based on 100% sequence homology), mouse (based on 100% sequence homology), rat (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer228)

Gene Information

human ... CARM1(10498)

General description

CARM1 (coactivator-associated arginine methyltransferase 1) is an arginine N-methyltransferase that catalyzes the transfer of a methyl group from S-adenosyl-L-methionine to arginine residues. This protein-arginine methyltransferase that works in conjunction with p300 and GRIP1 to enhance transcription. CARM1 methylation targets include histone H3 at Arg17 and p300 at Arg2142. CARM1 is recruited to promoters upon gene activation together with histone acetyltransferases from the p300 and p160 families. During monocyte inflammatory stimulation, CARM1 acts together with p300 as a coactivator for NF-kB. It is responsible for moving cells toward the inner cell mass in developing blastocysts. CARM1 also plays an important role in androgen receptors and may play a role in prostate cancer progression.

Specificity

This antibody recognizes CARM1 phosphorylated at Ser228.

Immunogen

Epitope: Phosphorylated Ser228
KLH-conjugated linear peptide corresponding to human CARM1 phosphorylated at Ser228.

Application

Anti-phospho CARM1 (Ser228) Antibody is a Rabbit Polyclonal Antibody for detection of phospho CARM1 (Ser228) also known as Protein arginine N-methyltransferase 4 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Histones

Quality

Evaluated by Western Blot in EGF treated and untreated A431 cell lysates.

Western Blot Analysis: 2 µg/mL of this antibody detected CARM1 on 10 µg of EGF treated and untreated A431 cell lysates.

Target description

~63 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
EGF treated and untreated A431 cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service