Skip to Content
Merck
All Photos(2)

Key Documents

ABC821

Sigma-Aldrich

Anti-AMBRA1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Activating molecule in BECN1-regulated autophagy protein 1, AMBRA1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AMBRA1(55626)

General description

Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. Beclin-1, a principal regulator of autophagosome formation (3), is in turn regulated by AMBRA1. AMBRA1 associates with Beclin-1 through a region near its center as determined by yeast two-hybrid assay. Null mutations in this gene in mice resulted in embryonic lethality with severe neural tube defects associated with autophagy impairment, accumulation of ubiquitinated proteins, unbalanced cell proliferation and excessive apoptotic death. Furthermore, down-regulation of AMBRA1 in cultured cells though RNA interference decreased the level of rapamycin and nutrient starvation-induced autophagy (4). Multiple isoforms of AMBRA1 are known to exist.

Immunogen

Epitope: N-terminus
KLH-conjugated synthetic peptide corresponding to the N-terminus of human AMBRA1.

Application

Anti-AMBRA1 Antibody is an antibody against AMBRA1 for use in western blotting, IHC & immunofluorescence.
Immunohistochemistry Analysis: 5 μg/mL from a representative lot detected AMBRA1 in human brain tissue.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected AMBRA1 in human brain cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: 2 μg/mL of this antibody detected AMBRA1 in rat brain tissue lysate.

Target description

~150 kDa observed. Uncharacterized bands may be observed in some lysate(s). Uniprot describes 6 isoforms produced by alternative splicing ranging between ~84 kDa and ~143 kDa

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Strobbe et al.
British journal of pharmacology, 178(2), 298-311 (2020-10-11)
The mitochondrial F1 Fo -ATPsynthase is pivotal for cellular homeostasis. When respiration is perturbed, its mode of action everts becoming an F1 Fo -ATPase and therefore consuming rather producing ATP. Such a reversion is an obvious target for pharmacological intervention

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service