Skip to Content
Merck
All Photos(1)

Documents

AB3042

Sigma-Aldrich

Anti-Uncoupling Protein 2 Antibody

Chemicon®, from rabbit

Synonym(s):

UCP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Ucp2(22228)

Specificity

Uncoupling Protein 2 (UCP2), mouse. Mouse UCP2 is a 309 amino acid mitochondrial uncoupling protein (Fleury et al. 1997). It is only 59% homologous with UCP1 found in brown adipose tissues. Mouse UCP2 (chromosome 7) has approximately 95% homology with human UCP2 located on chromosome 11 (Fleury et al. 1997). UCP2 has wide tissue distribution in mouse tissues (brain, kidney, liver, brown adipose tissue, heart, and muscle). UCP2 may play a critical role in energy balance, body weight, and thermoregulation (Fleury et al. 1997; Flier & Lowell 1997).

SPECIES REACTIVITIES: The antibody has not yet been tested for cross-reactivity to other species. The sequence used for immunization shows 92% homology to rat and 86% homology to human UCP2.

Immunogen

14 aa peptide sequence at the C-terminus of the mouse UCP2 (Fleury et al. 1997). The peptide has no significant homology with UCP1.

Application

Research Category
Metabolism
Research Sub Category
Ion & Transport Channels
This Anti-Uncoupling Protein 2 Antibody is validated for use in ELISA, WB for the detection of Uncoupling Protein 2.
Western Blotting: 1-10 μg/mL using Chemiluminescence technique.

ELISA: 1:10,000-1:100,000; using 50-100 ng UCP2 control peptide/well.

Optimal working dilutions must be determined by end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS with 0.1% BSA .

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristina A Simeone et al.
Experimental neurology, 251, 84-90 (2013-11-26)
Mitochondria actively participate in neurotransmission by providing energy (ATP) and maintaining normative concentrations of reactive oxygen species (ROS) in both presynaptic and postsynaptic elements. In human and animal epilepsies, ATP-producing respiratory rates driven by mitochondrial respiratory complex (MRC) I are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service