Skip to Content
Merck
All Photos(1)

Key Documents

9Q0718

Anti-Rotavirus A MAb Antibody, clone 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

antibody form

affinity purified immunoglobulin

clone

clone 1
monoclonal

purified by

affinity chromatography

species reactivity

all, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable
immunodiffusion: suitable
western blot: suitable

isotype

IgG2b

General description

Viral Lysate

Specificity

Specific for intact virus of strains: Rhesus Rotavirus, human rotavirus WA strain, SA11, etc.

Application

Recommended pair for double antibody immunoassay: Coating: MABX8452; labeling: MABX8451

Physical form

50 mM NaCl, 10 mM PBS pH 7.5, with 0.1% NaN3

Storage and Stability

2-8°C. For long-term storage, freeze at -20°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service