Skip to Content
Merck
All Photos(1)

Key Documents

8.52287

Sigma-Aldrich

Fmoc-p-Bz-Phe-OH

Fmoc-L-p-benzoylphenylalanine Novabiochem®

Synonym(s):

Fmoc-p-Bz-Phe-OH, (S)-3-(4-Benzoyl-phenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-propionic acid,4-Benzoyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]phenylalanine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€169.00
5 G
€667.00

€169.00


Estimated to ship onApril 12, 2025


Request a Bulk Order

Select a Size

Change View
1 G
€169.00
5 G
€667.00

About This Item

Empirical Formula (Hill Notation):
C31H25NO5
CAS Number:
Molecular Weight:
491.53
MDL number:
UNSPSC Code:
12352209

€169.00


Estimated to ship onApril 12, 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

N([C@@H](Cc4ccc(cc4)C(=O)c5ccccc5)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C31H25NO5/c33-29(21-8-2-1-3-9-21)22-16-14-20(15-17-22)18-28(30(34)35)32-31(36)37-19-27-25-12-6-4-10-23(25)24-11-5-7-13-26(24)27/h1-17,27-28H,18-19H2,(H,32,36)(H,34,35)/t28-/m0/s1

InChI key

SYOBJKCXNRQOGA-NDEPHWFRSA-N

General description

Fmoc-p-Bz-Phe-OH-OH is a useful tool for preparing photoactivatable peptide-based affinity probes [1]. On photolysis at 366 nm, Benzoylphenylalanine (Bpa) generates a biradical that has a preference for insertion into C-H bonds, particularly those of Leu, Val and Met side chains. The derivative can be introduced using standard coupling method such as PyBOP and is stable to conditions used in peptide chain extension. For the cleavage of Bpa containing peptide from the resin, the use of thiols and silanes should be avoided as dithioketal formation and reduction, respectively, have been observed.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] K. T. O′Neil, et al. (1989) J. Biol. Chem., 264.

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in DMF): -31.0 - -27.0 °
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service