Skip to Content
Merck
All Photos(1)

Key Documents

8.45066

Sigma-Aldrich

Sodium bromate

Msynthplus

Synonym(s):

Sodium bromate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BrNaO3
CAS Number:
Molecular Weight:
150.89
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-160-4

form

crystals

potency

400 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: oxidant

mp

381 °C (decomposition)

solubility

364 g/L

density

3.3 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/BrHO3.Na/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

XUXNAKZDHHEHPC-UHFFFAOYSA-M

Analysis Note

Colour (visual): white
Appearance of substance (visual): crystals
Assay (iodometric): ≥ 99.0 %
pH-value (10 % in water): 6.0 - 7.0
1,1-Dichloroethene (HS-GC): lot specific result
1,1,1-Trichloroethane (HS-GC): lot specific result
Carbon tetrachloride (HS-GC): lot specific result
Benzene (HS-GC): lot specific result
1,2-Dichloroethane (HS-GC): lot specific result

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 1 - Skin Irrit. 2

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service