Skip to Content
Merck
All Photos(1)

Documents

8.43984

Sigma-Aldrich

Phenol solution

for synthesis (90% solution in water)

Synonym(s):

Phenol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O
CAS Number:
Molecular Weight:
94.11
Beilstein:
6428463
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.22

product name

Phenol (90% solution in water), for synthesis

Quality Level

Assay

≥99% (GC)

form

liquid

transition temp

flash point 81 °C (Information refers to the main component.)

density

1.07 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenol, also known as hydroxybenzene, is a significant benzene derivative in commercial applications. It serves as a fundamental feedstock to produce polycarbonates, epoxy resins, molding compounds, and phenolic resins. Moreover, phenol finds extensive use in the manufacturing of aniline, caprolactam, and various other chemical derivatives.

Application

Phenol (90% solution in water) is used as a solvent to synthesis low molecular weight phenol-formaldehyde resins.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

177.8 °F - Information refers to the main ingredient.

Flash Point(C)

81 °C - Information refers to the main ingredient.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service