Skip to Content
Merck
All Photos(1)

Key Documents

8.41200

Sigma-Aldrich

2-Chloro-6-fluorobenzoic acid

for synthesis

Synonym(s):

2-Chloro-6-fluorobenzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€201.00

€201.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

Select a Size

Change View
25 G
€201.00

About This Item

CAS Number:
Molecular Weight:
174.55
MDL number:
UNSPSC Code:
12352106
EC Index Number:
207-105-2
NACRES:
NA.22

€201.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

Quality Level

form

powder

mp

156-159 °C

storage temp.

2-30°C

SMILES string

Fc1c(c(ccc1)Cl)C(=O)O

InChI

1S/C7H4ClFO2/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,(H,10,11)

InChI key

XNTIGDVFBDJLTQ-UHFFFAOYSA-N

Related Categories

General description

for synthesis

Application

2-Chloro-6-fluorobenzoic acid for synthesis. CAS 434-75-3, molar mass 174.55 g/mol.

Analysis Note

Assay (acidimetric)≥ 98.0 %Melting range (lower value)≥ 156 °CMelting range (upper value)≤ 159 °CIdentity (IR)passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service