Skip to Content
Merck
All Photos(1)

Key Documents

8.21050

Sigma-Aldrich

2-Pyrazinecarboxamide

for synthesis

Synonym(s):

2-Pyrazinecarboxamide, Pyrazine-2-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€49.60

€49.60


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
25 G
€49.60

About This Item

Empirical Formula (Hill Notation):
C5H5N3O
CAS Number:
Molecular Weight:
123.11
MDL number:
UNSPSC Code:
12352111
EC Index Number:
202-717-6
NACRES:
NA.22

€49.60


Estimated to ship onMay 26, 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

form

powder

pH

7 ( in H2O)

mp

188-191 °C

storage temp.

2-30°C

SMILES string

NC(=O)c1nccnc1

InChI

1S/C5H5N3O/c6-5(9)4-3-7-1-2-8-4/h1-3H,(H2,6,9)

InChI key

IPEHBUMCGVEMRF-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 188 °C
Melting range (upper value): ≤ 191 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service