Skip to Content
Merck
All Photos(1)

Documents

8.18660

Sigma-Aldrich

N-Benzyl-tert-butylamine

for synthesis

Synonym(s):

N-Benzyl-tert-butylamine, N-tert-Butylbenzylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NHC(CH3)3
CAS Number:
Molecular Weight:
163.26
UNSPSC Code:
12352116
EC Index Number:
222-179-6
NACRES:
NA.22

vapor pressure

0.07 hPa ( 53 °C)

Quality Level

Assay

≥97.0% (GC)

potency

400 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

mp

-30--25 °C

transition temp

flash point 90 °C

solubility

2 g/L

density

0.90 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C11H17N/c1-11(2,3)12-9-10-7-5-4-6-8-10/h4-8,12H,9H2,1-3H3/p+1

InChI key

DLSOILHAKCBARI-UHFFFAOYSA-O

Related Categories

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.895 - 0.897
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

194.0 °F

Flash Point(C)

90 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service