Skip to Content
Merck
All Photos(1)

Documents

8.18556

Sigma-Aldrich

2-Cyclohexylethanol

for synthesis

Synonym(s):

2-Cyclohexylethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O
CAS Number:
Molecular Weight:
128.21
MDL number:
UNSPSC Code:
12352104
EC Index Number:
224-672-1

vapor pressure

0.2 hPa ( 25 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

potency

940 mg/kg LD50, oral (Rat)
1220 mg/kg LD50, skin (Rabbit)

expl. lim.

0.9-6.3 % (v/v)

mp

-20 °C

transition temp

flash point 93 °C

density

0.92 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H16O/c9-7-6-8-4-2-1-3-5-8/h8-9H,1-7H2

InChI key

QJQZRLXDLORINA-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.920 - 0.922
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service