Skip to Content
Merck
All Photos(1)

Documents

8.18382

Sigma-Aldrich

Diethylene glycol mono-n-hexyl ether

for synthesis

Synonym(s):

Diethylene glycol mono-n-hexyl ether, 2-(2-n-Hexyloxyethoxy)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)2O(CH2)2O(CH2)5CH3
CAS Number:
Molecular Weight:
190.28
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-988-3

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

210 °C

potency

2400 mg/kg LD50, oral (Rat)
1395 mg/kg LD50, skin (Rabbit)

expl. lim.

1.1-6.3 % (v/v)

pH

5-7 (20 °C, 200 g/L in H2O)

bp

260 °C/1013 hPa

mp

-41--39 °C

transition temp

flash point 123 °C

density

0.93 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C10H22O3/c1-2-3-4-5-7-12-9-10-13-8-6-11/h11H,2-10H2,1H3

InChI key

GZMAAYIALGURDQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.932 - 0.934
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

253.4 °F

Flash Point(C)

123 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service