Skip to Content
Merck
All Photos(3)

Key Documents

8.16016

Sigma-Aldrich

(S)-(+)-Glutamine

for synthesis

Synonym(s):

L-Glutamine, L-Glutamic acid-5-amide, Gln, (S)-2,5-Diamino-5-oxopentanoic acid, L-Glutamic acid 5-amide, Levoglutamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€31.20
100 G
€85.80

€31.20


Only 0 left in stock

Request a Bulk Order

Select a Size

Change View
25 G
€31.20
100 G
€85.80

About This Item

Linear Formula:
H2NCOCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
146.14
Beilstein:
1723797
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-292-1
NACRES:
NA.22

€31.20


Only 0 left in stock

Request a Bulk Order

Quality Level

form

solid

potency

7500 mg/kg LD50, oral (Rat)

pH

4.0-6.0 (20 °C, 25 g/L in H2O)

mp

185 °C (dec.) (lit.)

solubility

100 g/L

density

1.47 g/cm3 at 20 °C

bulk density

640 kg/m3

storage temp.

2-30°C

SMILES string

N[C@@H](CCC(N)=O)C(O)=O

InChI

1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m0/s1

InChI key

ZDXPYRJPNDTMRX-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

L-Glutamine is an essential amino acid that is a crucial component of culture media that serves as a major energy source for cells in culture. L-Glutamine is very stable as a dry powder and as a frozen solution. In liquid media or stock solutions, however, L-glutamine degrades relatively rapidly. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use.

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Spec. rotation [α²0/D (c=10 in HCl 2 mol/l): +31.5 - +33.0 °
Identity (IR): passes test

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service