Skip to Content
Merck
All Photos(2)

Key Documents

8.08626

Sigma-Aldrich

3,3′-Thiodipropionic acid

for synthesis

Synonym(s):

3,3′-Thiodipropionic acid, Thiodihydracrylic acid, Bis(2-carboxyethyl)sulfide

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€42.70

€42.70


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
250 G
€42.70

About This Item

Linear Formula:
S(CH2CH2COOH)2
CAS Number:
Molecular Weight:
178.21
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-841-3
NACRES:
NA.22

€42.70


Estimated to ship onMay 21, 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (acidimetric)

form

powder

potency

3000 mg/kg LD50, oral (Rat)

mp

128-131 °C

transition temp

flash point 126-129 °C

storage temp.

2-30°C

SMILES string

S(CCC(=O)O)CCC(=O)O

InChI

1S/C6H10O4S/c7-5(8)1-3-11-4-2-6(9)10/h1-4H2,(H,7,8)(H,9,10)

InChI key

ODJQKYXPKWQWNK-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Melting range (lower value): ≥ 128 °C
Melting range (upper value): ≤ 131 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

262.4 °F - closed cup

Flash Point(C)

128 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service