Skip to Content
Merck
All Photos(1)

Key Documents

8.03521

Sigma-Aldrich

Diallylamine

for synthesis

Synonym(s):

Diallylamine, N-2-Propenyl-2-propen-1-amine

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€56.00
1 L
€138.00

€56.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€56.00
1 L
€138.00

About This Item

Linear Formula:
(CH2=CHCH2)2NH
CAS Number:
Molecular Weight:
97.16
MDL number:
UNSPSC Code:
12352116
EC Index Number:
204-671-2
NACRES:
NA.22

€56.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

vapor pressure

24.2 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

273 °C

potency

578 mg/kg LD50, oral (Rat)
>500 mg/kg LD50, skin (Rabbit)

expl. lim.

0.93-14.2 % (v/v)

pH

11.5 (9.7 g/L in H2O)

bp

111 °C/1013 hPa

mp

-88 °C

transition temp

flash point 11 °C

solubility

86 g/L

density

0.79 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CC=C)CC=C

InChI

1S/C6H11N/c1-3-5-7-6-4-2/h3-4,7H,1-2,5-6H2

InChI key

DYUWTXWIYMHBQS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.787 - 0.792
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service