Skip to Content
Merck
All Photos(1)

Key Documents

73021BC

Millipore

GDS OTV Insert

BioControl, For use with GDS, Performance verification of the GDS Rotor-Gene

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€639.00

€639.00


Estimated to ship onMay 24, 2025


Request more information

Select a Size

Change View
1 EACH
€639.00

About This Item

UNSPSC Code:
41116135

€639.00


Estimated to ship onMay 24, 2025


Request more information

Quality Level

shelf life

1‑5 mo.

manufacturer/tradename

BioControl Systems 73021

storage condition

protect from light at room temperature

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS Rotor-Gene

storage temp.

room temp

General description

The GDS OTV (Optical Temperature Verification) Insert provides automated performance verification and dynamic calibration of the GDS Rotor-Gene. The OTV system uses the chemical properties of thermochromic liquid crystals (TLC) as an absolute temperature reference. When heated, a TLC changes from opaque to transparent at a very precise temperature. The Rotor-Gene can measure this precise temperature transition point and, by comparing the measured in-tube thermal transition point to the known calibrated value, the OTV software verifies that the Rotor-Gene is within specification. If necessary, the OTV software allows the user to automatically recalibrate the instrument with a simple push of a button. A printable post-run report documents each test and any necessary recalibration.

Components

  • OTV Insert – good for 30 verification runs
  • Optical Filters for Rotor-Gene 3000 and Rotor-Gene Q, one each

Testing Method

Optical Temperature Verification

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service