Skip to Content
Merck
All Photos(1)

Documents

666125

Sigma-Aldrich

Urea Transporter B Inhibitor, UTBinh-14

Synonym(s):

Urea Transporter B Inhibitor, UTBinh-14, 3-(4-Ethylphenylsulfonyl)-N-(thiophen-2-ylmethyl)thieno[2,3-e][1,2,3]triazolo[1,5-a]pyrimidin-5-amine, UT-B Inhibitor, UTB Inhibitor, UTBinh14, 3-((4-Ethylphenyl)sulfonyl)-N-(2-thienylmethyl)thieno[2,3-e][1,2,3]triazolo[1,5-a]pyrimidin-5-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17N5O2S3
Molecular Weight:
455.58
UNSPSC Code:
12352200
NACRES:
NA.28

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable triazolothienopyrimidine compound that acts as a reversible urea transporter B (UT-B)-selective inhibitor (IC50 = 10.3 and 25.1 nM against urea influx using human and mouse erythrocytes, respectively) by targeting a UT-B intracellular site in a urea-competitive manner, exhibiting little effect toward UT-A1, UT-A3, AQP1, CFTR, or TMEM16A channel activities and no cytotoxicity in MDCK cultures (100% viability after 24 h 10 µM inhibitor treatment). Shown to effectively decrease maximum urinary concentration (2.3- and 3.7-fold of basal [urea], respectively, with or without inhibitor; 300 µg/300 µl/mouse i.p.) and increase urination volume (236 vs.109 µl/4h, respectively, with or without inhibitor) from antidiuretic dDAVP- (1 µg/kg; administered 1 h after inhibitor) treated wild-type, but not UT-B-/-, mice in vivo. Despite its short half-life (t1/2 = 2.8 min) in in vitro rat hepatic microsomes stability tests, an average inhibitor concentration of 390 µM in blood can be reached in mice even 6 h after a single i.p. dose of 200 µl per animal.
A cell-permeable triazolothienopyrimidine compound that acts as a reversible urea transporter B-selective inhibitor (IC50 = 10.3 and 25.1 nM against urea influx using human and mouse erythrocytes, respectively) by targeting a UT-B intracellular site in a urea-competitive manner, exhibiting little effect toward UT-A1, UT-A3, AQP1, CFTR, or TMEM16A channel activities. Shown to effectively decrease maximum urinary concentration and increase urination volume from antidiuretic dDAVP-treated wild-type, but not UT-B-/-, mice (300 µg/300 µl/mouse i.p.) in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
urea transporter B
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Anderson, M.O., et al. 2012. J. Med. Chem.55, 5942.
Yao, C., et al. 2012. J. Am. Soc. Nephrol.23, 1210.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service