Skip to Content
Merck
All Photos(1)

Key Documents

584222

Millipore

Tetrahydrouridine

Potent competitive inhibitor of cytidine deaminase. Also available as a 100 mM solution in H2O.

Synonym(s):

Tetrahydrouridine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€272.00

€272.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€272.00

About This Item

Empirical Formula (Hill Notation):
C9H16N2O6
CAS Number:
Molecular Weight:
248.23
MDL number:
UNSPSC Code:
12352208
NACRES:
NA.51

€272.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Quality Level

description

RTECS - UW5798980

Assay

≥90% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white to off-white

solubility

water: 200 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

N2(CCC(NC2=O)O)[C@@H]1O[C@@H]([C@H]([C@H]1O)O)CO

InChI

1S/C9H16N2O6/c12-3-4-6(14)7(15)8(17-4)11-2-1-5(13)10-9(11)16/h4-8,12-15H,1-3H2,(H,10,16)/t4-,5?,6-,7-,8-/m1/s1

InChI key

UCKYOOZPSJFJIZ-XVKVHKPRSA-N

General description

A potent competitive inhibitor of cytidine deaminase. Used in combination with cytosine arabinoside (Ara-C) to assess the anti-leukemic activity and anti-tumor activity of Ara-C in in vitro studies.

Application


  • Oncotherapy resistance explained by Darwinian and Lamarckian models.: This research explores the mechanisms of oncotherapy resistance, combining Darwinian and Lamarckian models to provide a comprehensive understanding. The study highlights the role of Tetrahydrouridine in modulating therapy resistance pathways in cancer treatment (Saunthararajah et al., 2024).

  • Neuroendocrine lineage commitment of small cell lung cancers can be leveraged into p53-independent non-cytotoxic therapy.: This study investigates the potential of using Tetrahydrouridine in non-cytotoxic therapies for small cell lung cancers, focusing on neuroendocrine lineage commitment and its implications for treatment efficacy (Biswas et al., 2023).

  • In Vitro Interaction of Tetrahydrouridine with Key Human Nucleoside Transporters.: This study explores how Tetrahydrouridine interacts with human nucleoside transporters, shedding light on its mechanisms of action and potential as a therapeutic agent in biochemistry and pharmacology (Säll et al., 2023).

  • Mycoplasma infection of cancer cells enhances anti-tumor effect of oxidized methylcytidines.: This research investigates how mycoplasma infection enhances the anti-tumor effects of oxidized methylcytidines, with Tetrahydrouridine playing a crucial role in the observed therapeutic outcomes (Pang et al., 2023).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot, purge with inert gas, and freeze (-20°C). Aqueous stock solutions (at neutral or basic pH) are stable for up to 3 months at -20°C.

Other Notes

Yusa, K., et al. 1995. Biochem. Biophys. Res. Commun. 206, 486.
Bouffard, D.Y., et al. 1993. Biochem Pharmacol.45, 1857.
Laliberte, J., et al. 1992. Cancer Chemotherap. Pharmacol.30, 7.
Riva, C., et al. 1992. Chemotherapy38, 358.
Yusa, K., et al. 1992. J. Biol. Chem. 267, 16848.
Hanze, A.R. 1967. J. Am. Chem. Soc.89, 6720.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. Dear partener, Kindly advice if THU 10 mg vials sored at -80 for 3 days can be used. Thank you,

    1 answer
    1. The stability of this material at -80°C has not been determined. In solid form, compounds are generally less sensitive to temperature, particularly for relatively short periods of time. It is likely that the quality has not been impacted. However this cannot be guaranteed. It would be up to the end user to determine the suitability for use.

      Helpful?

  2. Hi, may I ask if the THU (Cat number: 18771-50-1) is stable if stored at -4 oC and 10 oC? There is temperature excursion of the freezer in lab, and we would like to check what is the highest tolerable storage Temp which this reagent stored

    1 answer
    1. The recommended storage temperature for this material is −20°C. The product is shipped on ice to maintain the quality of the compound. In general, products of this nature are considered stable within the range of −10 to 25°C. The end user would have to evaluate the integrity of the product for excursions beyond this range.

      Helpful?

  3. Does this product have an expiry date?

    1 answer
    1. This product does not contain an expiration date. It contains a recommended retest date of 10 years after the quality release date. A recommended retest date is the period of time during which the product is expected to remain within established stability specifications, provided that it has been stored under defined conditions. After the Retest Date, product samples should be examined to ensure that the product is still in compliance with the established specifications. For more information, you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management/quality-services

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service