Skip to Content
Merck
All Photos(2)

Key Documents

557353

Sigma-Aldrich

RORγ Inverse Agonist, SR2211

The RORγ Inverse Agonist, SR2211 controls the biological activity of RORγ.

Synonym(s):

RORγ Inverse Agonist, SR2211, 1,1,1,3,3,3-hexafluoro-2-(2-fluoro-4′-((4-(pyridin-4-ylmethyl)piperazin-1-yl)methyl)-[1,1′-biphenyl]-4-yl)propan-2-ol, SR-2211, SR 2211

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H24F7N3O
CAS Number:
Molecular Weight:
527.48
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥99% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC1=CC(C(O)(C(F)(F)F)C(F)(F)F)=CC=C1C2=CC=C(CN3CCN(CC4=CC=NC=C4)CC3)C=C2

General description

A cell-permeable, piperazine containing biphenyl compound that binds directly to retinoic acid receptor related orphan receptor γ (RORg) and acts as a highly selective and inverse agonist (Ki = 105 nM; IC50 ~ 320 nM). Reported to block the transcriptional activity of RORg and suppress the synthesis of IL-17 in EL-4 murine lymphoma cell line. Exhibits only a minimal effect on RORa and LXRa activity.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kumar, N., et al. 2012. ACS Chem. Biol.7, 672.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuzhu Hou et al.
Protein & cell, 9(12), 1027-1038 (2018-03-07)
Macrophages acquire distinct phenotypes during tissue stress and inflammatory responses. Macrophages are roughly categorized into two different subsets named inflammatory M1 and anti-inflammatory M2 macrophages. We herein identified a unique pathogenic macrophage subpopulation driven by IL-23 with a distinct gene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service