Skip to Content
Merck
All Photos(2)

Key Documents

554726

Sigma-Aldrich

SR9009

≥96% (HPLC), liquid, REV-ERB agonist, Calbiochem®

Synonym(s):

REV-ERB Agonist II, SR9009, Ethyl-3-(((4-chlorobenzyl)((5-nitrothiophen-2-yl)methyl)amino)methyl)pyrrolidine-1-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€242.00

€242.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€242.00

About This Item

Empirical Formula (Hill Notation):
C20H24ClN3O4S
CAS Number:
Molecular Weight:
437.94
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€242.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

Product Name

REV-ERB Agonist II, SR9009, The REV-ERB Agonist II, SR9009 controls the biological activity of REV-ERB. This small molecule/inhibitor is primarily used for Biochemicals applications.

Quality Level

Assay

≥96% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark brown

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

ClC(C=C1)=CC=C1CN(CC2=CC=C(S2)[N+]([O-])=O)CC3CN(C(OCC)=O)CC3

InChI

1S/C20H24ClN3O4S/c1-2-28-20(25)23-10-9-16(13-23)12-22(11-15-3-5-17(21)6-4-15)14-18-7-8-19(29-18)24(26)27/h3-8,16H,2,9-14H2,1H3

InChI key

MMJJNHOIVCGAAP-UHFFFAOYSA-N

General description

A pyrrolidinecarbamate compound that acts as a specific REV-ERB-α/β agonist. Exhibits a direct and reversible binding (Kd = 800 nM REV-ERB-α) and shows excellent selectivity over a panel of 46 other nuclear receptors. Potently increases REV-ERB-dependent repressor activity (IC50 = 670 and 800 nM in Gal4-REV-ERB-α and β transfected HEK 293 cells) and reduces BMAL1 mRNA in HepG2 cells (IC50 = 710 nM). Shown to cross the blood-brain barrier and exhibit desirable pharmacokinetic properties. A 7-day administration of SR9009 (100 mg/kg, i.p., b.i.d.) causes a significant weight and fat loss in C57BL6 mice with concomitant reduction in plasma glucose and free fatty acid levels, and alters circadian pattern.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
REV-ERB-α/&beta
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Solt, L.A., et al. 2012. Nature485, 62.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaodong Zhuang et al.
iScience, 24(10), 103144-103144 (2021-09-22)
The coronavirus disease 2019 pandemic, caused by severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) coronavirus, is a global health issue with unprecedented challenges for public health. SARS-CoV-2 primarily infects cells of the respiratory tract via spike glycoprotein binding to angiotensin-converting
Fang-Yuan Mao et al.
Cellular and molecular gastroenterology and hepatology, 12(2), 395-425 (2021-03-07)
Rev-erbα represents a powerful transcriptional repressor involved in immunity. However, the regulation, function, and clinical relevance of Rev-erbα in Helicobacter pylori infection are presently unknown. Rev-erbα was examined in gastric samples from H pylori-infected patients and mice. Gastric epithelial cells (GECs)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service