Pseudosubstrate region of the ε-isotype of protein kinase C (PKCε) with an alanine to serine substitution. PKCε exhibits an apparent specificity for this peptide (Km = 68 µM).
Biochem/physiol Actions
Km = 68 µM for PKCε
Primary Target Pseudosubstrate region of the ε-isotype of protein kinase C (PKCε)
Following initial thaw, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Schaap, D. and Parker, P.J. 1990. J. Biol. Chem. 265, 7301. Schaap, D., et al. 1989. FEBS Lett. 243, 351.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.