Skip to Content
Merck
All Photos(1)

Key Documents

5.30310

Sigma-Aldrich

Mithramycin A

Synonym(s):

Mithramycin A, Plicamycin, Mithramycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H76O24
CAS Number:
Molecular Weight:
1085.15
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

InChI

1S/C52H76O24/c1-18-29(72-34-14-30(43(58)21(4)68-34)73-33-13-28(54)42(57)20(3)67-33)12-26-10-25-11-27(49(66-9)48(63)41(56)19(2)53)50(47(62)39(25)46(61)38(26)40(18)55)76-36-16-31(44(59)23(6)70-36)74-35-15-32(45(60)22(5)69-35)75-37-17-52(8,65)51(64)24(7)71-37/h10,12,19-24,27-28,30-37,41-45,49-51,53-61,64-65H,11,13-17H2,1-9H3/t19-,20-,21-,22-,23-,24-,27+,28-,30-,31-,32-,33+,34+,35+,36+,37+,41+,42-,43-,44-,45-,49+,50+,51-,52+/m1/s1

InChI key

CFCUWKMKBJTWLW-WZIVUMCRSA-N

General description

An aureolic acid type tricyclic pentaglycoside antibiotics with anti-tumor properties. Binds to G-C rich DNA and displaces Sp1 transcription factor from its sites in the promoters of selected oncogenes, such as c-Myc and c-Src. Mithramycin A-induced inhibition of Sp1 activity has been linked to greater DNA damage and cell death in cells that are homozygous for SNP309. Specifically reduces the levels of MDM2 in human ductal breast epithelial tumor cell line, T47D (5-fold reduction at 200 nM), but does not affect cells that are wild-type for SNP309. Shown to induce myeloid differentiation of leukemic blasts that leads to reduction in the level of expression of c-Myc and c-Abl proto oncogenes that harbor G-C rich promoters. Inhibits Mcl-1 activity and mTOR phosphorylation (Ser2448) in prostate cancer cells without affecting normal human prostate epithelial cells. Shown to reduce expression of endogenous transient receptor potential vanilpod receptor (TRPV1)-like protein in dorsal root ganglionic neurons.

Biochem/physiol Actions

Primary Target
Sp1 transcription factor
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zavala, K., et al. 2014. Neurosci Lett.562,in press.
Choi, E. S., et al. 2013. J. Clin. Biochem. Nutr.53, 89.

Sleiman S. F., et al. 2011. J. Neurosci.31, 6858.
Bond, G, et al. 2004. Cell.119, 591.
Blume, S, et al. 1991. J. Clin. Invest.88, 1613.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service