Skip to Content
Merck
All Photos(1)

Key Documents

5.05984

Sigma-Aldrich

Mitochondrial Content Inducer, BRD6897

Synonym(s):

Mitochondrial Content Inducer, BRD6897, BRD 6897, 5,6-Dimethyl-2-(2-(2-methyl-1H-indol-3-yl)-2-oxoethylthio)-3-phenylthieno[2,3-d]pyrimidin-4(3H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H21N3O2S2
CAS Number:
Molecular Weight:
459.58
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

General description

A cell-permeable indolylphenylthienopyrimidinone compound that increases mitochondrial content without affecting the cell size (~10 µM). Reduces mitochondrial protein turnover without affecting any known transcriptional programs of mitochondrial biogenesis. Reversibly increases uncoupled respiration in non-dividing cells (~50% increase in 3T3-L1 and HUVEC at 10 µM), but appears to be toxic to dividing cells. Shown to increase the expression of mitochondrial DNA encoded oxidative phosphorylation (OxPhos) genes without affecting nuclear DNA encoded OxPhos gene expression.

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Kitami, T., et al. 2012. PLoS ONE7, e33755.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service