Skip to Content
Merck
All Photos(1)

Key Documents

5.05298

Sigma-Aldrich

p300/CBP Inhibitor VI, L002

Synonym(s):

p300/CBP Inhibitor VI, L002, 4-(4-Methoxyphenylsulfonyloxyimino)-2,6-dimethyl-2,5-cyclohexadien-1-one, GCN5 Inhibitor III, Histone Acetyltransferase Inhibitor IX, PCAF Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15NO5S
CAS Number:
Molecular Weight:
321.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brown

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(ON=C2C=C(C(=O)C(=C2)C)C)c1ccc(cc1)OC

InChI

1S/C15H15NO5S/c1-10-8-12(9-11(2)15(10)17)16-21-22(18,19)14-6-4-13(20-3)5-7-14/h4-9H,1-3H3

InChI key

VEWFTYOFWIXCIO-UHFFFAOYSA-N

General description

A cell-permeable phenylsulfonyloxyiminocyclohexadienone compound that acts as a potent, selective, and reversible inhibitor of p300 histone acetyl transferase (IC50 = 1.98 µM). Shown to occupy Ac-CoA binding pocket of p300, and less potent to CBP, PCAF, GCN5 and GNAT. Suppresses Histone H3 and H4 acetylation in triple negative breast cancer cell line MDAMB-468 as well as in HCT116 cell line and blocks p300-mediated STAT3 phosphorylation in pancreatic cancer cell line MIA Paca-2. Does neither inhibit the activities of histone deacetylases 1, 6, and 11 nor 8-diverse histone methyltransferases tested (DOT1, EZH1, G9a, PRMT1, SETD2, SET7-9, SMYD2, and SUV39H2). Reported to suppress tumor growth in mice bearing tumor xenografts of MDAMB-468 cell line (at 500 µg/100 µl, i.p. twice a week).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
p300 HAT
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Yang, H., et al. 2013. Mol. Cancer Ther.12, 610.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service