Skip to Content
Merck
All Photos(1)

Documents

5.04972

Sigma-Aldrich

ATR Inhibitor IV

Synonym(s):

ATR Inhibitor IV, VE821, 3-amino-6-(4-(methylsulfonyl)phenyl)-N-phenylpyrazine-2-carboxamide, VE-821, ATM and Rad3-Related Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N4O3S
CAS Number:
Molecular Weight:
368.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

13 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

InChI

1S/C18H16N4O3S/c1-26(24,25)14-9-7-12(8-10-14)15-11-20-17(19)16(22-15)18(23)21-13-5-3-2-4-6-13/h2-11H,1H3,(H2,19,20)(H,21,23)

InChI key

DUIHHZKTCSNTGM-UHFFFAOYSA-N

General description

A cell-permeable phenylpyrazine derivative that acts as a potent, ATP-competitive (Ki= 13 nM; IC50/[ATP] = 70 nM/50 µM and 128 nM/100 µM), ATR-selective inhibitor with much reduced or little potency against 4 related PIKKs (Ki= 2.2, 3.9, 16, and >1 µM, respectively, against DNA-PK, PI 3-Kγ, ATM, and mTOR) and 50 other kinases. Shown to selectively inhibit ATR-dependent H2AX Ser139 & Chk1 Ser345 phosphorylation (complete inhibition at 10 µM in HT29 and HFL1 cultures) without affecting ATM- and DNA-PK-mediated H2AX or ATM-mediated Chk2 phosphorylation. Short-term (24 h) VE-821 treatment, either alone or in synergy with Cisplatin (Cat. No. 232120), results in reversible cytostatic growth arrest regardless of cellular ATM-p53 pathway activity, while cytotoxicity and Cisplatin synergism in cell death induction is reported to occur only upon long-term VE-821 exposure (≥72 h) in cultures with ATM-p53 pathway defects.
A cell-permeable phenylpyrazine derivative that acts as a potent, ATP-competitive (Ki= 13 nM; IC50/[ATP] = 70 nM/50 µM and 128 nM/100 µM), ATR-selective inhibitor with much reduced or little potency against 4 related PIKKs (Ki= 2.2, 3.9, 16, and >1 µM, respectively, against DNA-PK, PI 3-Kγ, ATM, and mTOR) and 50 other kinases. Shown to selectively inhibit ATR-dependent H2AX Ser139 & Chk1 Ser345 phosphorylation (complete inhibition at 10 µM in HT29 and HFL1 cultures) without affecting ATM- and DNA-PK-mediated H2AX or ATM-mediated Chk2 phosphorylation. Short-term (24 h) VE-821 treatment, either alone or in synergy with Cisplatin (Cat. No. 232120), results in reversible cytostatic growth arrest regardless of cellular ATM-p53 pathway activity, while cytotoxicity and Cisplatin synergism in cell death induction is reported to occur only upon long-term VE-821 exposure (≥72 h) in cultures with ATM-p53 pathway defects.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Prevo, R., et al. 2012, Cancer Biol. Ther.13, 1072; Pires, I.M., et al. 2012, Br. J. Cancer107, 291; Reaper, P.M., et al. 2011, Nat. Chem. Biol.7, 428.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service