Skip to Content
Merck
All Photos(1)

Key Documents

489475

Sigma-Aldrich

Anti-LRP-5/6 Mouse mAb (1A12)

liquid, clone 1A12, Calbiochem®

Synonym(s):

Anti-Low Density Lipoprotein Receptor-Related Protein-5/6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

1A12, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

mouse, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2b

shipped in

wet ice

storage temp.

−70°C

Gene Information

human ... LRP5(4041)

General description

Purified mouse monoclonal antibody. Recognizes the ~175 kDa full length LRP-5/6 and the ~145 kDa proteolytic fragment.
Recognizes the ~175 kDa full-length LRP-6 protein and a ~145 kDa proteolytic fragment.
This Anti-LRP-5/6 Mouse mAb (1A12) is validated for use in Immunoblotting for the detection of LRP-5/6.

Immunogen

Human
a synthetic peptide (DTGTDRIEVTR) corresponding to amino acids 421-431 of the human low density lipoprotein receptor-related protein 6 (LRP-6)

Application

Immunoblotting (1-2 µg/ml)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 10 mM Tris-glycine, 25% glycerol, 0.5% BSA, pH 7.0.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Li, L., et al. 2001. J. Biol. Chem.277, 5977.
Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service