Skip to Content
Merck
All Photos(1)

Key Documents

360570-M

Sigma-Aldrich

Pyridine

ACS reagent, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N
CAS Number:
Molecular Weight:
79.10
Beilstein:
103233
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
Assay:
≥99.0%
bp:
115 °C (lit.)
vapor pressure:
10 mmHg ( 13.2 °C)
20 mmHg ( 25 °C)
Pricing and availability is not currently available.

grade

ACS reagent

vapor density

2.72 (vs air)

vapor pressure

10 mmHg ( 13.2 °C)
20 mmHg ( 25 °C)

Assay

≥99.0%

autoignition temp.

899 °F

expl. lim.

12.4 %

impurities

Reducing substances, passes test
≤0.002% NH3
0.1% water

evapn. residue

≤0.002%

refractive index

n20/D 1.509 (lit.)

bp

115 °C (lit.)

mp

−42 °C (lit.)

solubility

water: soluble

density

0.978 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.001%

cation traces

Cu: ≤5 ppm

SMILES string

C1=CN=CC=C1

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H

InChI key

JUJWROOIHBZHMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyridine (Pyr) is a nitrogen containing six membered heterocyclic compound commonly used as a base. It is a colorless liquid with a fishy odor. Pyridine moiety is present as a major component in a number of compounds like pyridine nucleotides,[1] pyridine alkaloids,[2] pyridine-based polymers[3] and pyridine-containing macrocycles.[4] It has wide application as a solvent, catalyst and as an intermediate for synthesis.[5] IR, Raman and UV spectra of pyridine has been analyzed and the values obtained were used to calculate its thermodynamic properties.[6] Photodegradation[7] and bacterial degradation of Pyr has been studied.[8]

Application

Pyridine may be used in the synthesis of the following:
  • complexes with cobalt(II) halides[9]
  • pyridine-d5, a deuterated form of pyridine[10]
  • ionic liquid matrices (ILMs)[11]

Other Notes

Important notice
This product will be phased out towards early 2019. There is a replacement product available : 360570

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jasmeet Kaur et al.
Environmental science and pollution research international, 20(6), 3956-3964 (2013-05-08)
This study demonstrates the influences of position, number of nitrogen (N) atoms and -C-N- or -N=N- linkage present in the six membered heterocyclic compounds such as pyridine, pyrazine, and pyridazine on their photocatalytic degradation by Au, Ag, and Fe(+2) deposited
R A Pandey et al.
Bioresource technology, 98(12), 2258-2267 (2006-07-04)
Industrial waste gas emissions containing pyridine are generated from pyridine manufacturing industries, and in industrial operations where pyridine is used as a solvent, as an intermediate for synthesis and as a catalyst for a variety of applications. Pyridine has unpleasant
Hani Nasser Abdelhamid et al.
Analytica chimica acta, 767, 104-111 (2013-03-05)
Application of two new series of ionic liquid matrices (ILMs) based on the two most predominantly used conventional organic matrices (Sinapinic acid and 2,5-DHB) in conjugation with various bases (aniline (ANI), dimethyl aniline (DMANI), diethylamine (DEA), dicyclohexylamine (DCHA), pyridine (Pyr)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service