Skip to Content
Merck
All Photos(1)

Key Documents

324904

Sigma-Aldrich

Epiregulin, Mouse, Recombinant

Epiregulin, Mouse, Recombinant is expressed in E. coli with an N-terminal methionine. Inhibits epithelial tumor growth and stimulates the growth of fibroblasts and other cell types.

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€564.00

€564.00


Estimated to ship onMay 05, 2025



Select a Size

Change View
50 μG
€564.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€564.00


Estimated to ship onMay 05, 2025


recombinant

expressed in E. coli

Quality Level

Assay

≥97% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<1.0 EU/μg protein Endotoxin

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, mouse epiregulin expressed in E. coli with an N-terminal methionine. Epiregulin is shown to inhibit epithelial tumor growth, able to stimulate growth of fibroblasts and other cell types, and has been shown to be an autocrine growth factor in human epidermal keratinocytes.

Biochem/physiol Actions

ED₅₀ = 0.3-1 ng/ml as measured in a cell proliferation assay using Balb/3T3 mouse embryonic fibroblasts.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.2 µm sterile-filtered PBS, BSA.

Reconstitution

Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C.
Reconstitute to 10 µg/ml in sterile PBS containing at least 0.1% human or bovine serum albumin.

Other Notes

Shirakata, Y., et al. 2000. J. Biol. Chem.275, 5748.
Shelly, M., et al. 1998. J. Biol. Chem.273, 10496.
Das, S.K., et al. 1997. Dev. Biol.190, 178.
Komurasaki, T., et al. 1997. Oncogene15, 2841.
Toyoda, H., et al. 1995. J. Biol. Chem.270, 7495.
Toyoda, H., et al. 1995. FEBS Lett.377, 403.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service