Skip to Content
Merck
All Photos(1)

Key Documents

251800

Sigma-Aldrich

Daunorubicin, Hydrochloride

Potent cell-permeable anticancer agent whose potential target site may be mitochondrial cytochrome c oxidase.

Synonym(s):

Daunorubicin, Hydrochloride, Daunomycin, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29NO10 · xHCl
CAS Number:
Molecular Weight:
527.52 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

form

crystalline solid

potency

≥840 g/kg

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

red

solubility

aqueous buffer: soluble
methanol: soluble
water: soluble

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C27H29NO10.ClH/c1-10-22(30)14(28)7-17(37-10)38-16-9-27(35,11(2)29)8-13-19(16)26(34)21-20(24(13)32)23(31)12-5-4-6-15(36-3)18(12)25(21)33;/h4-6,10,14,16-17,22,30,32,34-35H,7-9,28H2,1-3H3;1H/t10-,14-,16-,17-,22+,27-;/m0./s1

InChI key

GUGHGUXZJWAIAS-QQYBVWGSSA-N

General description

Potent cell-permeable anticancer agent whose potential target site may be mitochondrial cytochrome C oxidase. Has been shown to inhibit RNA and DNA synthesis and to intercalate into DNA. Inhibits eukaryotic topoisomerase I and II. Induces DNA single strand breaks. Induces apoptosis in HeLaS3 tumor cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
DNA synthesis, RNA synthesis
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Aqueous stock solutions are stable for up to 1 month at 4°C.

Other Notes

Friedenberg, W.R., et al. 1995. Am. J. Clin. Oncol. 18, 105.
Papadopoulou, L.C., and Tsiftsoglou, A.S. 1993. Cancer Res. 53, 1072.
Skladanowski, A., and Konopa, J. 1993. Biochem. Pharmacol.46, 375.
Wolff, M., and Jelkmann, W. 1993. Ann. Hematol. 66, 27.
Bellec, F., et al. 1992. Flow Cytometry 13, 880.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Muta. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pietri Puustinen et al.
Autophagy, 16(10), 1871-1888 (2020-01-28)
Macroautophagy/autophagy is a central component of the cytoprotective cellular stress response. To enlighten stress-induced autophagy signaling, we screened a human kinome siRNA library for regulators of autophagic flux in MCF7 human breast carcinoma cells and identified the catalytic subunit of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service