Skip to Content
Merck
All Photos(1)

Key Documents

207000

Sigma-Aldrich

Calcineurin Autoinhibitory Peptide

The Calcineurin Autoinhibitory Peptide controls the biological activity of Mn2+-stimulated calcineurin activity. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Calcineurin Autoinhibitory Peptide, ITSFEEAKGLDRINERMPPRRDAMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C124H205N39O39S2
Molecular Weight:
2930.32
UNSPSC Code:
12352202

Quality Level

Assay

≥97% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: soluble

shipped in

ambient

storage temp.

−20°C

General description

Corresponds to a C-terminal domain (residues 457-482) of the calmodulin-binding domain of calcineurin. Specific calcineurin inhibitor. Does not inhibit CaM kinase II, protein phosphatase I or 2A. Inhibits Mn2+-stimulated calcineurin activity (IC50 = 10 µM using 32P myosin light chain as a substrate) but has no effect on Ni2+-stimulated activity.
Specific calcineurin inhibitor. Corresponds to the C-terminal domain (residues 457-482) of the calmodulin-binding domain of calcineurin (PP2B). Inhibits Mn2+-stimulated calcineurin activity (IC50 = 10 µM using 32P-myosin light chain as a substrate) but has no effect on Ni2+-stimulated enzyme activity. Does not inhibit CaM kinase II, protein phosphatase 1 or 2A.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Mn2+-stimulated calcineurin activity
Product does not compete with ATP.
Reversible: no
Target IC50: 10 µM using 32P-myosin light chain as a substrate

Packaging

Yes

Warning

Toxicity: Standard Handling (A)

Sequence

H-Ile-Thr-Ser-Phe-Glu-Glu-Ala-Lys-Gly-Leu-Asp-Arg-Ile-Asn-Glu-Arg-Met-Pro-Pro-Arg-Arg-Asp-Ala-Met-Pro-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Aqueous stock solutions are stable for up to 2 months at -20°C.

Other Notes

Perrino, B.A., et al. 1995. J. Biol. Chem. 270, 340.
Yokoyama, Y., and Wang, J.H. 1994. FEBS Lett.337, 128.
Yokoyama, N., et al. 1993. Arch. Biochem. Biophys.300, 615.
Matsui, H., et al. 1991. Biochem. Int.24, 1119.
Hashimoto, Y., et al. 1990. J. Biol. Chem.265, 1924.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service