Skip to Content
Merck
All Photos(1)

Key Documents

204884

Sigma-Aldrich

Complement C3a des-Arg, Human

Complement C3a des-Arg, Human, is a single chain native des-Arg C3 complement component

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€457.00

€457.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€457.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

€457.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Native, human des-Arg C3 complement component. Single chain, 76 amino acid peptide. Once the C3a peptide has been produced in human plasma or serum, it is rapidly converted to C3a des-Arg upon removal of the C-terminal arginine by endogenous serum carboxypeptidase N. This enzymatic process is considered to be a major mechanism for controlling C3a function in vivo because C3a des-Arg has <1% of the biological activity expressed by the C3a peptide. Therefore, C3a des-Arg can serve as a negative control molecule in experiments involved with the biological activities of the intact C3a anaphylatoxin peptide.

Biochem/physiol Actions

Ileum contraction assay: negative

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, pH 7.2.

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Kajita, T. and Hugli, T.E. 1991. Am. J. Pathol. 138, 1359.
Bokish, V.A. and Muller-Eberhard, H.J. 1970. J. Clin. Invest.49, 2427.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service