1.52371
Chromolith® WP 300 RP-18
L × I.D. 50 mm × 2 mm HPLC column
Synonym(s):
Chromolith® WP 300 RP-18 50-2 HPLC column
About This Item
Recommended Products
Quality Level
feature
endcapped: no
extent of labeling
18% loading
parameter
200 bar pressure
60 °C max. temp.
technique(s)
HPLC: suitable
mass spectrometry (MS): suitable
column L × I.D.
50 mm × 2 mm
matrix
monolithic silica
matrix active group
C18 bonding phase
pore size
2 μm macroporosity
30 mm mesoporosity
operating pH range
2-7.5
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The Chromolith® WP 300 RP-18 column gives unsurpassed RP-HPLC resolution of peptide maps from enzymatic digests. The improvements in silica and bonded-phase chemistry incorporated into the Chromolith® WP 300 line improve resolution by increasing efficiency and reducing peak tailing.
Chromolith® WP 300 columns are made of monolithic silica with a characteristic bimodal pore structure.
Chromolith® WP 300 RP-18 2 mm I.D. columns have macropores with 2 μm diameter, giving a column efficiency exceeding 80,000 plates/meter. The mesopores are 30 nm (300 Å), the surface modification is Octadecylsilane. Chromolith® WP 300 RP-18 columns fall under the USP L1 Classification. Chromolith® WP 300 RP-18 columns are fully suitable for LC-MS applications.
The column dimension of 50 mm length and 2 mm I.D. is the best choice for very fast separations of Biomolecules on any UHPLC System.
Chromolith® WP 300 RP-18 50-2 HPLC columns enable:
—Completely bioinert column hardware and high biorecovery
—High-speed separations at very low column back-pressure
—High matrix tolerance, allowing time and cost savings with simplified workflows
—Cost-savings due to extended column lifetimes
—Connection of columns, flow gradients and the use of mobile phases with higher viscosity
The recommended Guard-columns are:
Chromolith® WP 300 RP-18, 5-2 guard cartridges (3 pieces) [1.52372] with the Chromolith® Guard Cartridge Holder Bioinert 5 mm x 2 mm [1.52355]
The Chromolith® column coupler can be used to significantly increase the separation efficiency of Chromolith® columns by coupling several columns in series. [[1.51467]
Learn more:
Brochure: Race through your Separations on Any System. Chromolith® Monolithic Silica HPLC Columns
All Supelco HPLC Columns, including monolithic Chromolith® columns, are fully compatible to all HPLC and UHPLC Instruments.
Supelco HPLC Products: The Best Choice for Any LC Instrument
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Analytical Chromolith® HPLC columns for Bio applications, 300A mesopores: RP-18, RP-8, RP-4, Prot-A, Epoxy
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service