N-terminal 6×His-tagged Drosophila NAP1 expressed in E. coli.
Nucleosome assembly proteins (NAP) are involved in moving histones into the nucleus, nucleosome assembly and chromatin fluidity. They affect the transcription of many genes.
Application
Lot # R0803D0002: 12.5 µg NAP1 has been successfully used for 1 chromatin assembly reaction with Chromatin Assembly Kit (Millipore Corp, Catalog# 17-410).
Quality
Activity is routinely tested with Chromatin Assembly Kit (Millipore Corp, Catalog# 17-410).
Physical form
10 mM K-HEPES pH 7.6, 10 mM KCl, 50 mM NaCl, 0.1 mM EDTA, 10%(v/v) glycerol, 0.01% NP-40, 0.2 mM PMSF, 1mM DTT. Frozen solution.
Format: Purified
Storage and Stability
Stable for 6 months at -80°C from date of shipment. For maximum recovery of product, centrifuge original vial prior to removing the cap.
Other Notes
Specific Activity: Varies with lot
Varies with individual lot. Lot R0803D0002 has purity 90% by SDS-PAGE and Coomassie blue staining
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The study aimed to investigate the antibacterial activity of Mustard (Brassica juncea) and Moringa (Moringa oleifera) leaf extracts and coagulant protein for their potential application in water treatment. Bacterial cell aggregation and growth kinetics studies were employed for thirteen bacterial
We report data from single molecule studies on the interaction between single DNA molecules and core histones using custom-designed horizontal magnetic tweezers. The DNA-core histone complexes were formed using λ-DNA tethers, core histones, and NAP1 and were exposed to forces
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.