Skip to Content
Merck
All Photos(1)

Key Documents

1468400100

Millipore

Water

Purified, pH 5-7, suitable for microbiological culture, USP 71

Synonym(s):

Water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:
UNSPSC Code:
12191602
technique(s):
microbiological culture: suitable
application(s):
food and beverages
pharmaceutical
bp:
100 °C (lit.)
Pricing and availability is not currently available.

Product Name

Purified Water, pH 5.0-7.0

Agency

USP 71

Quality Level

feature

ready-to-use

technique(s)

microbiological culture: suitable

impurities

<1000 cfu/mL bacteria

refractive index

n20/D 1.34 (lit.)

pH

5.0-7.0

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

application(s)

food and beverages
pharmaceutical

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Purified Water is the solvent derived from non-portable water to meet the specifications described by regulatory authorities. The low microbial counts and pH, synergistically support the usage of purified water in pharma and food industries.

Application

Purified Water is mainly used for microbial media preparations and bulk material for non-parenteral formulations.
Apart from above, purified water finds its application in:
  • Cleaning of non-parenteral production equipment′s.
  • Input for WFI production.
  • Non-sterile tests and assays.
  • Preparation of Bulk Chemicals.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service