1.46804
Vegetable Peptone Agar – 90mm ICR+ Settle Plates
with Lecithin, Tween®, Histidine and sodium thiosulfate for environmental monitoring (Isolator and Clean room)- 30 mL
Synonym(s):
Vegetable Peptone Agar with neutralizers
About This Item
material
polystyrene plate
Quality Level
Agency
non-compendial
sterility
sterile; irradiated
form
plate (ready to use culture media)
solid
shelf life
12 mo.
feature
ready-to-use
composition
Vegetable Peptone Agar neutralizers: Lecithin, Polysorbate 80, Histidine and Sodium Thiosulfate
packaging
triple packed of
diam.
90 mm
pH
7.1-7.5
application(s)
air monitoring
environmental
pharmaceutical
compatibility
for use with MAS-100®
storage temp.
15-25°C
suitability
bacteria
molds
yeasts
General description
Application
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service