Skip to Content
Merck
All Photos(1)

Key Documents

14-525-M

Sigma-Aldrich

PKC ζ Protein, active, 10 µg

Active, N-terminal His6-tagged recombinant human PKC zeta, residues 2-end, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€547.00

€547.00


Estimated to ship onMay 18, 2025



Select a Size

Change View
10 μG
€547.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€547.00


Estimated to ship onMay 18, 2025


biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 68.9 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

General description

N-terminal His6-tagged recombinant human PKC zeta, residues 2-end
Product Source: Expressed by baculovirus in Sf21 insect cells

Application

Research Category
Metabolism

Inflammation & Immunology
Research Sub Category
Obesity

Metabolic Disorders

Osteoporosis

Arthritis

Biochem/physiol Actions

Protein Target: PKCζ
Target Sub-Family: AGC

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-525M when ordering the 250 µg size.

Quality

routinely evaluated by phosphorylation of PKC substrate peptide II

Physical form

Ni2+/NTA-agarose

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Birendra Kumar Shrestha et al.
The Journal of biological chemistry, 295(5), 1240-1260 (2019-12-21)
Human ATG8 family proteins (ATG8s) are active in all steps of the macroautophagy pathway, and their lipidation is essential for autophagosome formation. Lipidated ATG8s anchored to the outer surface of the phagophore serve as scaffolds for binding of other core

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service