1.15941
Light green SF yellowish (C.I. 42095)
for microscopy Certistain®
Synonym(s):
Light green SF yellowish (C.I. 42095)
About This Item
Recommended Products
Quality Level
form
solid
potency
>2000 mg/kg LD50, oral (Rat)
composition
Dye, ≥75% spectrophotometric assay
pH
2.9 (25 °C, 10 g/L in H2O)
solubility
200 g/L
bulk density
200 kg/m3
εmax
830-1130 at 629-634 nm in water at 0.005 g/L
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
2-30°C
SMILES string
[Na+].[Na+].[S](=O)(=O)([O-])c1cc(ccc1)CN(CC)c2ccc(cc2)[C+](c5ccc(cc5)[S](=O)(=O)[O-])c3ccc(cc3)N(CC)Cc4cc(ccc4)[S](=O)(=O)[O-]
InChI
1S/C37H36N2O9S3.2Na/c1-3-38(25-27-7-5-9-35(23-27)50(43,44)45)32-17-11-29(12-18-32)37(31-15-21-34(22-16-31)49(40,41)42)30-13-19-33(20-14-30)39(4-2)26-28-8-6-10-36(24-28)51(46,47)48;;/h5-24H,3-4,25-26H2,1-2H3,(H2-,40,41,42,43,44,45,46,47,48);;/q;2*+1/p-2
InChI key
DGOBMKYRQHEFGQ-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
As Certistain® dye it is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.
Analysis Note
Identity
- sodium:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (water): 629 - 634 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; water): 830 - 1130
TLC-Test: passes test
Loss on drying (110 °C): ≤ 12 %
Suitability for microscopy: passes test
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service