Skip to Content
Merck
All Photos(1)

Key Documents

1.09604

Supelco

n-Heptadecane

reference substance for gas chromatography

Synonym(s):

n-Heptadecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15CH3
CAS Number:
Molecular Weight:
240.47
MDL number:
UNSPSC Code:
12352001
EC Index Number:
211-108-4

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥99.3% (GC)

form

liquid

technique(s)

gas chromatography (GC): suitable

kinematic viscosity

<7 cSt(40 °C)

bp

302 °C/1013 hPa

mp

22 °C

transition temp

flash point 148 °C

density

0.78 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C17H36/c1-3-5-7-9-11-13-15-17-16-14-12-10-8-6-4-2/h3-17H2,1-2H3

InChI key

NDJKXXJCMXVBJW-UHFFFAOYSA-N

Analysis Note

Assay (GC): ≥ 99.3 %
Identity (IR-spectrum): conforms

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

300.2 °F - closed cup

Flash Point(C)

149 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service