Skip to Content
Merck
All Photos(1)

Key Documents

1.09082

Millipore

Adapter for glass Petri dish

For use with portable MAS-100® family

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€122.00

€122.00


Estimated to ship onMay 21, 2025


Request more information

Select a Size

Change View
1 UNIT
€122.00

About This Item

UNSPSC Code:
41103403
NACRES:
NA.74

€122.00


Estimated to ship onMay 21, 2025


Request more information

material

aluminum adapter

Agency

non-compendial

feature

autoclavable

packaging

pkg of 1 kit

manufacturer/tradename

MAS-100 NT®

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with MAS-100 Eco®
for use with MAS-100 NT®

storage temp.

no temp limit

General description

Adapter for glass Petri dish is an additional accessory for MAS-100® air samplers. The portable MAS-100® air samplers are designed to efficiently monitor air microbiology in cleanrooms and other controlled environments in which microbiological contamination has a direct impact on product quality, shelf life and deterioration. The adapter is padded with rubber discs to hold glass petri dishes in place without damaging them. It permits holding glass petri dishes in place on MAS-100®, MAS-100 NT®, MAS-100 NT® Ex and MAS-100 Eco® air samplers.

Application

Adapter for glass Petri dish is an additional accessory to hold glass Petri dishes in MAS-100® air samplers. Microbial air sampling devices are extensively used in Pharmaceutical and Personal care industries as well as Food, Beverage and Environmental applications.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 Eco is a registered trademark of MBV AG
MAS-100 NT is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service