Skip to Content
Merck
All Photos(2)

Documents

09-860

Sigma-Aldrich

Anti-DDX3X Antibody

serum, from rabbit

Synonym(s):

ATP-dependent RNA helicase DDX3X, DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, X-linked, DEAD box protein 3, X-chromosomal, DEAD box, X isoform, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3, DEAD/H box-3, Helicase-like protein 2, helicase like protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DDX3X(1654)

General description

DDX3X, DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, X-linked, is a member of the DEAD box family of proteins. Characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), DEAD box proteins are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. DDX3X is an ATP-dependent RNA helicase and acts as a co-factor for XPO1-mediated nuclear export of incompletely spliced HIV-1 Rev RNAs. DDX3X is also involved in HIV-1 replication and interacts specifically with hepatitis C virus core protein resulting in a change in intracellular location.

Specificity

This antibody detects RNA-binding protein DDX3X.

Immunogen

KLH-conjugated synthetic linear peptide corresponding to DDX3X.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Binding Protein (RBP)
Use Anti-DDX3X Antibody (Rabbit Polyclonal Antibody) validated in WB to detect DDX3X also known as ATP-dependent RNA helicase DDX3X, DEAD box protein 3 X-chromosomal.

Quality

Evaluated by western blot on A431 cell lysate.

Western Blot Analysis: 1:1000 dilution of this antibody detected DDX3X in A431 cell lysate.

Target description

~73 kDa

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
A431 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Severin Lechner et al.
Nature communications, 14(1), 3548-3548 (2023-06-16)
Lipoic acid is an essential enzyme cofactor in central metabolic pathways. Due to its claimed antioxidant properties, racemic (R/S)-lipoic acid is used as a food supplement but is also investigated as a pharmaceutical in over 180 clinical trials covering a
Adi Reches et al.
Journal of immunology (Baltimore, Md. : 1950), 196(12), 4967-4976 (2016-05-20)
MHC class I molecules, in addition to their role in specific activation of the CTL of adaptive immune system, function also as the main ligands for NK cell inhibitory receptors, which prevent NK cells from killing normal, healthy cells. MHC
Pietro Vella et al.
Nucleic acids research, 40(8), 3403-3418 (2012-01-03)
The Yin Yang 1 (YY1) transcription factor is a master regulator of development, essential for early embryogenesis and adult tissues formation. YY1 is the mammalian orthologue of Pleiohomeotic, one of the transcription factors that binds Polycomb DNA response elements in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service