Skip to Content
Merck
All Photos(1)

Key Documents

05-812

Sigma-Aldrich

Anti-NY-ESO-1 Antibody, clone E978

ascites fluid, clone E978, Upstate®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€660.00

€660.00


Estimated to ship onApril 14, 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1540

Request a Bulk Order

Select a Size

Change View
100 μL
€660.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€660.00


Estimated to ship onApril 14, 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1540

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

E978, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... CTAG1A(246100)

Specificity

NY-ESO-1

Immunogen

NY-ESO-1 full-length recombinant protein

Application

Anti-NY-ESO-1 Antibody, clone E978 detects level of NY-ESO-1 & has been published & validated for use in WB.
Research Category
Apoptosis & Cancer
Research Sub Category
Tumor Markers

Quality

routinely evaluated by immunoblot on RIPA lysates from SKMEL-19 cells

Target description

22kDa

Physical form

Ascites
mouse ascites with 0.05% sodium azide

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Stockert et al.
The Journal of experimental medicine, 187(8), 1349-1354 (1998-05-23)
Evidence is growing for both humoral and cellular immune recognition of human tumor antigens. Antibodies with specificity for antigens initially recognized by cytotoxic T lymphocytes (CTLs), e.g., MAGE and tyrosinase, have been detected in melanoma patient sera, and CTLs with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service